2-Brom-6-(4-morpholino)pyridin
Lieferant: COMBI-BLOCKS
Synonyme:
2-Bromo-6-morpholinopyridine
HC-6366-5G
HC-6366-25G
HC-6366-1G
COBBHC-6366-5GEA
153
EUR
COBBHC-6366-5G
COBBHC-6366-25G
COBBHC-6366-1G
2-Brom-6-(4-morpholino)pyridin
2-Brom-6-(4-morpholino)pyridin
Formel:
C₉H₁₁BrN₂O Molecular Weight: 243,1004 g/mol |
MDL:
MFCD07772824 CAS-Nummer: 332134-60-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...