N-Benzyl-2-brom-5-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4447-5G CA-4447-1G
COBBCA-4447-5GEA 451 EUR
COBBCA-4447-5G COBBCA-4447-1G
N-Benzyl-2-brom-5-fluorbenzamid
N-Benzyl-2-brom-5-fluorbenzamid
Formel: C₁₄H₁₁BrFNO
MDL: MFCD09800916
CAS-Nummer: 951884-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR