4-Brom-N-cyclopropyl-3-methoxybenzamid

Lieferant: COMBI-BLOCKS

CA-4646-5G CA-4646-1G
COBBCA-4646-5GEA 595 EUR
COBBCA-4646-5G COBBCA-4646-1G
4-Brom-N-cyclopropyl-3-methoxybenzamid
4-Brom-N-cyclopropyl-3-methoxybenzamid
Formel: C₁₁H₁₂BrNO₂
MDL: MFCD11504870
CAS-Nummer: 1072944-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR