2-Brom-N-cyclopentyl-5-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4439-1G CA-4439-5G
COBBCA-4439-1GEA 149 EUR
COBBCA-4439-1G COBBCA-4439-5G
2-Brom-N-cyclopentyl-5-fluorbenzamid
2-Brom-N-cyclopentyl-5-fluorbenzamid
Formel: C₁₀H₉BrFNO
MDL: MFCD09800910
CAS-Nummer: 951884-13-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR