3-Brom-N-cyclopentyl-4-fluorbenzamid

Lieferant: COMBI-BLOCKS

CA-4612-5G CA-4612-25G CA-4612-1G
COBBCA-4612-5GEA 303 EUR
COBBCA-4612-5G COBBCA-4612-25G COBBCA-4612-1G
3-Brom-N-cyclopentyl-4-fluorbenzamid
3-Brom-N-cyclopentyl-4-fluorbenzamid
Formel: C₁₀H₉BrFNO
MDL: MFCD11053799
CAS-Nummer: 639858-63-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR