5-Brom-N-butyl-2-methoxybenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0311-1G OT-0311-5G
COBBOT-0311-1GEA 124 EUR
COBBOT-0311-1G COBBOT-0311-5G
5-Brom-N-butyl-2-methoxybenzolsulfonamid
5-Brom-N-butyl-2-methoxybenzolsulfonamid
Formel: C₁₁H₁₆BrNO₃S
MDL: MFCD08056377
CAS-Nummer: 871269-18-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR