2-Brom-N-butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

HC-6129-5G HC-6129-1G
COBBHC-6129-5GEA 451 EUR
COBBHC-6129-5G COBBHC-6129-1G
2-Brom-N-butylbenzolsulfonamid
2-Brom-N-butylbenzolsulfonamid
Formel: C₁₀H₁₄BrNO₂S
MDL: MFCD09800950
CAS-Nummer: 951885-17-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR