3-Brom-N-tert-butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

Synonyme: 3-Bromo-N-(tert-butyl)benzenesulphonamide

OT-0148-5G OT-0148-1G
COBBOT-0148-5GEA 451 EUR
COBBOT-0148-5G COBBOT-0148-1G
3-Brom-N-tert-butylbenzolsulfonamid
3-Brom-N-tert-butylbenzolsulfonamid
Formel: C₁₀H₁₄BrNO₂S
Molecular Weight: 292.1900 g/mol
MDL: MFCD07363824
CAS-Nummer: 308283-47-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR