3-Brom-N-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0140-1G OT-0140-5G
COBBOT-0140-1GEA 212 EUR
COBBOT-0140-1G COBBOT-0140-5G
3-Brom-N-methylbenzolsulfonamid
3-Brom-N-methylbenzolsulfonamid
Formel: C₇H₈BrNO₂S
Molecular Weight: 250,11 g/mol
MDL: MFCD07363818
CAS-Nummer: 153435-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR