4-Brom-N-tert-butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0086-5G OT-0086-1G
COBBOT-0086-5GEA 385 EUR
COBBOT-0086-5G COBBOT-0086-1G
4-Brom-N-tert-butylbenzolsulfonamid
4-Brom-N-tert-butylbenzolsulfonamid
Formel: C₁₀H₁₄BrNO₂S
MDL: MFCD00458267
CAS-Nummer: 93281-65-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR