4-Brom-N-butylbenzolsulfonamid

Lieferant: COMBI-BLOCKS

OT-0082-5G OT-0082-1G
COBBOT-0082-5GEA 204 EUR
COBBOT-0082-5G COBBOT-0082-1G
4-Brom-N-butylbenzolsulfonamid
4-Brom-N-butylbenzolsulfonamid
Formel: C₁₀H₁₄BrNO₂S
MDL: MFCD01215086
CAS-Nummer: 1984-28-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR