N-(4-Bromphenyl)benzolsulfonamid ≥97%

Lieferant: Thermo Scientific
Achtung

H55476.MD H55476.03
H55476.MDEA 116 EUR
H55476.MD H55476.03
N-(4-Bromphenyl)benzolsulfonamid ≥97%
N-(4-Bromphenyl)benzolsulfonamid

Purity: 97% CAS No.: 16468-97-6 Molecular Formula: C12H10BrNO2S Formula Weight: 312.18 MDL No.: MFCD00159375

Formel: C₁₂H₁₀BrNO₂S
Molecular Weight: 312,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00159375
CAS-Nummer: 16468-97-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR