Methylvanillat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4-hydroxy-3-methoxybenzoate
CA-4973-5G
CA-4973-25G
CA-4973-100G
COBBCA-4973-5GEA
26.2
EUR
COBBCA-4973-5G
COBBCA-4973-25G
COBBCA-4973-100G
Methylvanillat
Methylvanillat
Formel:
C₉H₁₀O₄ Molecular Weight: 182,18 g/mol |
MDL:
MFCD00008438 CAS-Nummer: 3943-74-6 EINECS: 223-525-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...