Methylvanillat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-hydroxy-3-methoxybenzoate

M-Clarity™ Qualitätsstufe = MQ100

138126-100G 138126-25G
SIAL138126-100GEA 411 EUR
SIAL138126-100G SIAL138126-25G
Methylvanillat, Sigma-Aldrich®
Methylvanillat
Formel: C₉H₁₀O₄
Molecular Weight: 182,18 g/mol
MDL: MFCD00008438
CAS-Nummer: 3943-74-6
EINECS: 223-525-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR