2-Phenyl-4-thiazolcarbonsäure ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Phenylthiazole-4-carboxylic acid
H54521.MD
H54521.03
H54521.06
H54521.MDEA
33.4
EUR
H54521.MD
H54521.03
H54521.06
2-Phenyl-4-thiazolcarbonsäure ≥97%
2-Phenyl-4-thiazolecarboxylic acid
Purity: 97%
CAS No.: 7113-10-2
Molecular Formula: C10H7NO2S
Formula Weight: 205.23
Melting Point: 148-152°
MDL No.: MFCD00141954
Formel:
C₁₀H₇NO₂S Molecular Weight: 205,23 g/mol Schmelzpunkt: 148…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00141954 CAS-Nummer: 7113-10-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...