2-Phenyl-4-thiazolcarbonsäure

Lieferant: BIONET RESEARCH

Synonyme: 2-Phenylthiazole-4-carboxylic acid

12F-317S.500 12F-317S.1 12F-317S.5 12F-317S.10
BIOT12F-317S.500EA 0 EUR
BIOT12F-317S.500 BIOT12F-317S.1 BIOT12F-317S.5 BIOT12F-317S.10
2-Phenyl-4-thiazolcarbonsäure
2-Phenyl-4-thiazolecarboxylic acid
Formel: C₁₀H₇NO₂S
Molecular Weight: 205.23 g/mol
MDL: MFCD00141954
CAS-Nummer: 7113-10-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR