2-Methyl-3-nitrophenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Methyl-3-nitrophenyl)boronic acid

BB-2474-5G BB-2474-1G
COBBBB-2474-5GEA 2230 EUR
COBBBB-2474-5G COBBBB-2474-1G
2-Methyl-3-nitrophenylboronsäure
2-Methyl-3-nitrophenylboronsäure
Formel: C₇H₈BNO₄
Molecular Weight: 180,9537 g/mol
MDL: MFCD06201025
CAS-Nummer: 1072945-60-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR