Cyclopentylboronsäure 95%

Lieferant: Thermo Fisher Scientific
Achtung

H27081.09 H27081.03
H27081.09EA 160 EUR
H27081.09 H27081.03
Cyclopentylboronsäure 95%
Cyclopentylboronic acid

Purity: 95% CAS No.: 63076-51-7 Molecular Formula: C5H11BO2 Formula Weight: 113.95 MDL No.: MFCD01074541

Formel: C₅H₁₁BO₂
Molecular Weight: 113.95 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01074541
CAS-Nummer: 63076-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR