Cyclopentylboronsäure

Lieferant: FLUOROCHEM

011025-5G 011025-1G
FLUO011025-5GEA 87.9 EUR
FLUO011025-5G FLUO011025-1G
Cyclopentylboronsäure
Cyclopentylboronic acid
Formel: C₅H₁₁BO₂
Molecular Weight: 113.95 g/mol
MDL: MFCD01074541
CAS-Nummer: 63076-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR