Cyclopentylboronsäure, Sigma-Aldrich®

Lieferant: Merck


MQ100

588415-10G 588415-1G
SIAL588415-10GEA 274 EUR
SIAL588415-10G SIAL588415-1G
Cyclopentylboronsäure, Sigma-Aldrich®
Cyclopentylboronic acid
Formel: C₅H₁₁BO₂
Molecular Weight: 113.95 g/mol
MDL: MFCD01074541
CAS-Nummer: 63076-51-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR