N-Fmoc-D-norleucin
Lieferant: BACHEM BIOCHEMICA
4027625.0005
4027625.0025
BACE4027625.0005EA
0
EUR
BACE4027625.0005
BACE4027625.0025
N-Fmoc-D-norleucin
N-Fmoc-D-norleucin
Formel:
C₂₁H₂₃NO₄ |
MDL:
MFCD00155639 CAS-Nummer: 112883-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...