N-Fmoc-L-norleucin
Lieferant: BACHEM BIOCHEMICA
Synonyme:
Fmoc-Nle-OH, FMOC-L-Norleucine
4004485.0005
4004485.0025
BACE4004485.0005EA
0
EUR
BACE4004485.0005
BACE4004485.0025
N-Fmoc-L-norleucin
N-Fmoc-L-norleucin
Formel:
C₂₁H₂₃NO₄ Molecular Weight: 353,4 g/mol |
MDL:
MFCD00037537 CAS-Nummer: 77284-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...