4-Brom-1H-pyrazol
Lieferant: COMBI-BLOCKS
HI-1701-5G
HI-1701-25G
HI-1701-100G
COBBHI-1701-5GEA
0
EUR
COBBHI-1701-5G
COBBHI-1701-25G
COBBHI-1701-100G
4-Brom-1H-pyrazol
4-Bromo-1H-pyrazole
Formel:
C₃H₃BrN₂ Molecular Weight: 146.97 g/mol |
MDL:
MFCD00075602 CAS-Nummer: 2075-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...