2-(Methoxycarbonyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(2-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Methoxycarbonylphenylboronic acid pinacol ester
PN-2125-5G
PN-2125-25G
PN-2125-1G
COBBPN-2125-5GEA
121
EUR
COBBPN-2125-5G
COBBPN-2125-25G
COBBPN-2125-1G
2-(Methoxycarbonyl)phenylboronsäurepinakolester
2-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₄H₁₉BO₄ Molecular Weight: 262,1093 g/mol |
MDL:
MFCD05663866 CAS-Nummer: 653589-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...