3-(Methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(3-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Methoxycarbonylphenylboronic acid pinacol ester

PN-2116-5G PN-2116-25G PN-2116-1G
COBBPN-2116-5GEA 0 EUR
COBBPN-2116-5G COBBPN-2116-25G COBBPN-2116-1G
3-(Methoxycarbonyl)phenylboronsäurepinakolester
3-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
MDL: MFCD03789258
CAS-Nummer: 480425-35-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR