4-(Methoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

BB-2108-25G BB-2108-5G
COBBBB-2108-25GEA 141 EUR
COBBBB-2108-25G COBBBB-2108-5G
4-(Methoxycarbonyl)phenylboronsäurepinakolester
4-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₄H₁₉BO₄
Molecular Weight: 262,11 g/mol
MDL: MFCD02179438
CAS-Nummer: 171364-80-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR