3-(2-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-3855-1G
BB-3855-5G
COBBBB-3855-1GEA
0
EUR
COBBBB-3855-1G
COBBBB-3855-5G
3-(2-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
3-(2-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
Formel:
C₁₄H₁₃BClNO₃ |
MDL:
MFCD09027260 CAS-Nummer: 957060-97-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...