3-(3-Chlor-2-methylphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5139-5G BB-5139-1G
COBBBB-5139-5GEA 303 EUR
COBBBB-5139-5G COBBBB-5139-1G
3-(3-Chlor-2-methylphenylcarbamoyl)phenylboronsäure
3-(3-Chlor-2-methylphenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₃BClNO₃
MDL: MFCD09972109
CAS-Nummer: 1072946-02-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR