4-(2-Chlor-4-methylphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((2-Chloro-4-methylphenyl)carbamoyl)phenyl)boronic acid

BB-3208-1G BB-3208-5G
COBBBB-3208-1GEA 186 EUR
COBBBB-3208-1G COBBBB-3208-5G
4-(2-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
4-(2-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₃BClNO₃
Molecular Weight: 289.52 g/mol
MDL: MFCD09027213
CAS-Nummer: 913835-39-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR