4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-(3-Chloro-4-methylphenylcarbamoyl)benzeneboronic acid

BB-3206-5G BB-3206-1G
COBBBB-3206-5GEA 277 EUR
COBBBB-3206-5G COBBBB-3206-1G
4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
4-(3-Chlor-4-methylphenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₃BClNO₃
Molecular Weight: 289.53 g/mol
MDL: MFCD09027211
CAS-Nummer: 913835-37-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR