4-(4-Chlorphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-((4-Chlorophenyl)carbamoyl)phenyl)boronic acid

BB-3123-5G BB-3123-1G
COBBBB-3123-5GEA 0 EUR
COBBBB-3123-5G COBBBB-3123-1G
4-(4-Chlorphenylcarbamoyl)phenylboronsäure
4-(4-Chlorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BClNO₃
Molecular Weight: 275.4953 g/mol
MDL: MFCD09027203
CAS-Nummer: 874288-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR