3-(3-Chlor-4-fluorphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-5141-5G BB-5141-1G
COBBBB-5141-5GEA 0 EUR
COBBBB-5141-5G COBBBB-5141-1G
3-(3-Chlor-4-fluorphenylcarbamoyl)phenylboronsäure
3-(3-Chlor-4-fluorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₀BClFNO₃
MDL: MFCD09972110
CAS-Nummer: 1072946-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR