5-(Diethylcarbamoyl)-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-(Diethylcarbamoyl)-2-fluorophenyl)boronic acid

BB-3342-5G BB-3342-1G
COBBBB-3342-5GEA 1180 EUR
COBBBB-3342-5G COBBBB-3342-1G
5-(Diethylcarbamoyl)-2-fluorphenylboronsäure
5-(Diethylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239.0500 g/mol
MDL: MFCD08436045
CAS-Nummer: 874289-47-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR