4-(tert-Butylcarbamoyl)-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(tert-Butylcarbamoyl)-3-fluorophenyl)boronic acid

BB-3314-5G BB-3314-1G
COBBBB-3314-5GEA 1490 EUR
COBBBB-3314-5G COBBBB-3314-1G
4-(tert-Butylcarbamoyl)-3-fluorphenylboronsäure
4-(tert-Butylcarbamoyl)-3-fluorphenylboronsäure
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239.0500 g/mol
MDL: MFCD08689525
CAS-Nummer: 874289-18-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR