3-Fluor-4-(isopropylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3312-5G BB-3312-1G
COBBBB-3312-5GEA 0 EUR
COBBBB-3312-5G COBBBB-3312-1G
3-Fluor-4-(isopropylcarbamoyl)phenylboronsäure
3-Fluor-4-(isopropylcarbamoyl)phenylboronsäure
Formel: C₁₀H₁₃BFNO₃
MDL: MFCD08436040
CAS-Nummer: 874289-16-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR