4-(Ethylcarbamoyl)-3-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Ethylcarbamoyl-3-fluorobenzeneboronic acid

BB-3308-5G BB-3308-1G
COBBBB-3308-5GEA 1250 EUR
COBBBB-3308-5G COBBBB-3308-1G
4-(Ethylcarbamoyl)-3-fluorphenylboronsäure
4-(Ethylcarbamoyl)-3-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Molecular Weight: 211.00 g/mol
MDL: MFCD08436070
CAS-Nummer: 874289-12-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR