5-(tert-Butylcarbamoyl)-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-(Tert-butylcarbmoyl)-2-fluorophenylboronic acid, 5-tert-Butylcarbamoyl-2-fluorobenzeneboronic acid, (5-(tert-Butylcarbamoyl)-2-fluorophenyl)boronic acid

BB-3346-5G BB-3346-1G
COBBBB-3346-5GEA 1580 EUR
COBBBB-3346-5G COBBBB-3346-1G
5-(tert-Butylcarbamoyl)-2-fluorphenylboronsäure
5-(tert-Butylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239,05 g/mol
MDL: MFCD08436054
CAS-Nummer: 874289-51-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR