5-(Butylcarbamoyl)-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-n-Butylcarbamoyl-2-fluorobenzeneboronic acid

BB-3345-5G BB-3345-1G
COBBBB-3345-5GEA 884 EUR
COBBBB-3345-5G COBBBB-3345-1G
5-(Butylcarbamoyl)-2-fluorphenylboronsäure
5-(Butylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₁₁H₁₅BFNO₃
Molecular Weight: 239,05 g/mol
MDL: MFCD08235104
CAS-Nummer: 874289-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR