5-(Ethylcarbamoyl)-2-fluorphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (5-(Ethylcarbamoyl)-2-fluorophenyl)boronic acid

BB-3340-5G BB-3340-1G
COBBBB-3340-5GEA 712 EUR
COBBBB-3340-5G COBBBB-3340-1G
5-(Ethylcarbamoyl)-2-fluorphenylboronsäure
5-(Ethylcarbamoyl)-2-fluorphenylboronsäure
Formel: C₉H₁₁BFNO₃
Molecular Weight: 211.0000 g/mol
MDL: MFCD08436027
CAS-Nummer: 874289-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR