2-Fluor-5-(propylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(2-Fluoro-5-(propylcarbamoyl)phenyl)boronic acid
BB-3343-1G
BB-3343-5G
COBBBB-3343-1GEA
0
EUR
COBBBB-3343-1G
COBBBB-3343-5G
2-Fluor-5-(propylcarbamoyl)phenylboronsäure
2-Fluor-5-(propylcarbamoyl)phenylboronsäure
Formel:
C₁₀H₁₃BFNO₃ |
MDL:
MFCD08436029 CAS-Nummer: 874289-48-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...