3-Fluor-4-(phenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3304-1G BB-3304-5G
COBBBB-3304-1GEA 0 EUR
COBBBB-3304-1G COBBBB-3304-5G
3-Fluor-4-(phenylcarbamoyl)phenylboronsäure
3-Fluor-4-(phenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
MDL: MFCD08436025
CAS-Nummer: 874288-40-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR