2-Fluor-5-(phenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Fluoro-5-(phenylcarbamoyl)phenyl)boronic acid

BB-3336-5G BB-3336-1G
COBBBB-3336-5GEA 0 EUR
COBBBB-3336-5G COBBBB-3336-1G
2-Fluor-5-(phenylcarbamoyl)phenylboronsäure
2-Fluor-5-(phenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
MDL: MFCD08235095
CAS-Nummer: 874289-41-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR