3-(2-Fluorphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3177-5G BB-3177-1G
COBBBB-3177-5GEA 0 EUR
COBBBB-3177-5G COBBBB-3177-1G
3-(2-Fluorphenylcarbamoyl)phenylboronsäure
3-(2-Fluorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
MDL: MFCD09027208
CAS-Nummer: 874288-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR