4-(2-(Trifluormethyl)phenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3211-5G BB-3211-1G
COBBBB-3211-5GEA 0 EUR
COBBBB-3211-5G COBBBB-3211-1G
4-(2-(Trifluormethyl)phenylcarbamoyl)phenylboronsäure
4-(2-(Trifluormethyl)phenylcarbamoyl)phenylboronsäure
Formel: C₁₄H₁₁BF₃NO₃
MDL: MFCD09027216
CAS-Nummer: 913835-42-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR