4-(2-Fluorphenylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-3127-1G
BB-3127-5G
COBBBB-3127-1GEA
0
EUR
COBBBB-3127-1G
COBBBB-3127-5G
4-(2-Fluorphenylcarbamoyl)phenylboronsäure
4-(2-Fluorphenylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₁BFNO₃ |
MDL:
MFCD09027205 CAS-Nummer: 874288-06-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...