4-(3-Fluorphenylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
4-(3-Fluorophenylcarbamoyl)benzeneboronic acid, (4-((3-Fluorophenyl)carbamoyl)phenyl)boronic acid
BB-3126-5G
BB-3126-1G
COBBBB-3126-5GEA
385
EUR
COBBBB-3126-5G
COBBBB-3126-1G
4-(3-Fluorphenylcarbamoyl)phenylboronsäure
4-(3-Fluorphenylcarbamoyl)phenylboronsäure
Formel:
C₁₃H₁₁BFNO₃ Molecular Weight: 259,04 g/mol |
MDL:
MFCD09027204 CAS-Nummer: 874288-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...