4-(4-Fluorphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3038-5G BB-3038-1G
COBBBB-3038-5GEA 0 EUR
COBBBB-3038-5G COBBBB-3038-1G
4-(4-Fluorphenylcarbamoyl)phenylboronsäure
4-(4-Fluorphenylcarbamoyl)phenylboronsäure
Formel: C₁₃H₁₁BFNO₃
MDL: MFCD06659878
CAS-Nummer: 850568-27-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR