Calcium-2-ethylhexanoat (3 - 8% Ca) in 2-Ethylhexansäure, Superleitende Qualität
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Ethylhexansäure Calciumsalz, Heptan-3-carbonsäure Calciumsalz, 2-Ethylcapronsäure Calciumsalz
36657.14
36657.06
36657.14EA
125
EUR
36657.14
36657.06
Calcium-2-ethylhexanoat (3 - 8% Ca) in 2-Ethylhexansäure, Superleitende Qualität
Calcium-2-ethylhexanoat
CAS No.: 136-51-6
Molecular Formula: C16H30CaO4
Formula Weight: 326.50
Physical Form: Liquid
Odour: Weak, characteristic
MDL No.: MFCD00014001
Formel:
C₁₆H₃₀CaO₄ Molecular Weight: 326,49 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014001 CAS-Nummer: 136-51-6 EINECS: 205-249-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...