Cer(III)oxalat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Cerium(III) oxalate hydrate
21122.06
21122.14
21122.22
21122.06EA
32.6
EUR
21122.06
21122.14
21122.22
Cer(III)oxalat Hydrat ≥99,9% (REO, Basis der Oxide der Metalle der seltenen Erden), REacton®
Cer(III)oxalat Hydrat
Purity: 99.9%
CAS No.: 15750-47-7
Molecular Formula: Ce2(C2O4)3.xH2O
Formula Weight: 544.29(anhy)
Physical Form: Powder
Sensitivity: Hygroscopic
Merck Reference: 14,2001
UN No.: 3288
MDL No.: MFCD00150139
Formel:
C₆Ce₂O₁₂·H₂O Molecular Weight: 544.29 g/mol (anhy) Schmelzpunkt: Zersetzung Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150139 CAS-Nummer: 15750-47-7 UN: 3288 ADR: 6.1 III Merck Index: 14,2001 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...