Erbium(III)oxalat Decahydrat 99.9%

Lieferant: Thermo Fisher Scientific
Achtung

A18420.06 A18420.14 A18420.22
A18420.06EA 21 EUR
A18420.06 A18420.14 A18420.22
Erbium(III)oxalat Decahydrat 99.9%
Erbium(III)oxalat Decahydrat

Purity: 99.9% CAS No.: 30618-31-6 Molecular Formula: C6Er2O12.10H2O Formula Weight: 778.73 (598.58anhy) UN No.: 3288 MDL No.: MFCD00150142

Formel: C₆Er₂O₁₂·10H₂O
Dichte: 2,64
Lagertemperatur: Raumtemperatur
MDL: MFCD00150142
CAS-Nummer: 30618-31-6
EINECS: 212-764-4
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR